Assay Details
Gene Symbol: IL22RA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034311.2 | NP_001029483.1 | 2 - 3 | 197 | 66 | |
GenBank mRNA | BC103413.1 | - | 2 - 3 | 198 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
