Assay Details
Gene Symbol: NIFK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034354.2 | NP_001029526.1 | 5 - 6 | 665 | 72 | |
GenBank mRNA | BC102791.1 | - | 5 - 6 | 668 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
