Assay Details
Gene Symbol: AOC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034361.2 | NP_001029533.1 | 4 - 5 | 2060 | 72 | |
GenBank mRNA | BC105152.1 | - | 4 - 5 | 2062 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
