Assay Details
Gene Symbol: L3HYPDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034386.1 | NP_001029558.1 | 1 - 2 | 590 | 76 | |
GenBank mRNA | BC104573.1 | - | 1 - 2 | 590 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
