Assay Details
Gene Symbol: PRRG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034402.2 | NP_001029574.1 | 5 - 6 | 585 | 100 | |
GenBank mRNA | BC102320.1 | - | 5 - 6 | 587 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
