Assay Details
Gene Symbol: PRRG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034402.2 | NP_001029574.1 | 6 - 7 | 723 | 97 | |
GenBank mRNA | BC102320.1 | - | 6 - 7 | 725 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
