Assay Details
Gene Symbol: CXCL14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034410.2 | NP_001029582.1 | 1 - 2 | 252 | 83 | |
GenBank mRNA | BC103335.1 | - | 1 - 2 | 253 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
