Assay Details
Gene Symbol: SBDS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034439.1 | NP_001029611.1 | 4 - 5 | 723 | 106 | |
GenBank mRNA | BC104585.1 | - | 4 - 5 | 723 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
