Assay Details
Gene Symbol: SAR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034521.2 | NP_001029693.1 | 1 - 2 | 103 | 80 | |
GenBank mRNA | BC102443.1 | - | 1 - 2 | 104 | 80 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC102443.1, NM_001034521.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
