Assay Details
Gene Symbol: BNIP3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034614.2 | NP_001029786.1 | 1 - 2 | 164 | 84 | |
GenBank mRNA | BC102612.1 | - | 1 - 2 | 165 | 84 | |
BT030544.1 | - | 1 - 2 | 160 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
