Assay Details
Gene Symbol: BNIP3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034614.2 | NP_001029786.1 | 3 - 4 | 417 | 72 | |
GenBank mRNA | BC102612.1 | - | 3 - 4 | 418 | 72 | |
BT030544.1 | - | 3 - 4 | 413 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
