Assay Details
Gene Symbol: BNIP3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034614.2 | NP_001029786.1 | 4 - 5 | 523 | 96 | |
GenBank mRNA | BC102612.1 | - | 4 - 5 | 524 | 96 | |
BT030544.1 | - | 4 - 5 | 519 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
