Assay Details
Gene Symbol: RPL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034625.2 | NP_001029797.1 | 1 | 26 | 62 | |
GenBank mRNA | BC102277.1 | - | 1 | 27 | 62 | |
BC151407.1 | - | 1 - 2 | 28 | 62 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC102277.1, BC151407.1, NM_001034625.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
