Assay Details
Gene Symbol: RPS10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034716.2 | NP_001029888.1 | 1 - 2 | 33 | 57 | |
GenBank mRNA | BC102416.1 | - | 1 - 2 | 36 | 57 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC102416.1, NM_001034716.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
