Assay Details
Gene Symbol: LAMTOR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034769.1 | NP_001029941.1 | 2 - 3 | 229 | 77 | |
GenBank mRNA | BC102442.1 | - | 2 - 3 | 229 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
