Assay Details
Gene Symbol: HMGN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034772.1 | NP_001029944.1 | 1 - 2 | 108 | 105 | |
GenBank mRNA | BC102449.1 | - | 1 - 2 | 108 | 105 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC102449.1, NM_001034772.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
