Assay Details
Gene Symbol: BCL7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034775.2 | NP_001029947.1 | 3 - 4 | 281 | 84 | |
GenBank mRNA | BC102475.1 | - | 3 - 4 | 282 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
