Assay Details
Gene Symbol: STMN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034790.2 | NP_001029962.1 | 1 - 2 | 28 | 74 | |
GenBank mRNA | BC102453.1 | - | 1 - 2 | 29 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC102453.1, NM_001034790.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
