Assay Details
Gene Symbol: GALM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034795.1 | NP_001029967.1 | 2 - 3 | 382 | 80 | |
GenBank mRNA | BC102446.1 | - | 2 - 3 | 382 | 80 | |
BT020690.1 | - | 2 - 3 | 399 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
