Assay Details
Gene Symbol: LOC616948 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034798.1 | NP_001029970.1 | 3 - 4 | 725 | 122 | |
GenBank mRNA | AY970972.1 | - | 3 - 4 | 725 | 122 | |
DQ381151.1 | - | 2 - 3 | 514 | 122 |
Gene Symbol: TRIM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205305.1 | NP_001192234.1 | 3 - 4 | 892 | 122 | |
GenBank mRNA | BC119899.1 | - | 3 - 4 | 892 | 122 | |
DQ380509.1 | - | 2 - 3 | 514 | 122 | ||
DQ381150.1 | - | 2 - 3 | 514 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
