Assay Details
Gene Symbol: MSRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034810.2 | NP_001029982.2 | 2 - 3 | 359 | 115 | |
GenBank mRNA | BC105188.1 | - | 2 - 3 | 307 | 115 | |
CK836798.1 | - | 2 - 3 | 359 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
