Assay Details
Gene Symbol: MTRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035013.2 | NP_001030185.1 | 4 - 5 | 674 | 146 | |
GenBank mRNA | BC105224.1 | - | 4 - 5 | 675 | 146 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
