Assay Details
Gene Symbol: MYL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035025.2 | NP_001030197.1 | 3 - 4 | 215 | 81 | |
GenBank mRNA | BC102914.1 | - | 3 - 4 | 216 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
