Assay Details
Gene Symbol: MGST3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035046.2 | NP_001030218.1 | 2 - 3 | 145 | 160 | |
GenBank mRNA | BC102190.1 | - | 2 - 3 | 146 | 160 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC102190.1, NM_001035046.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
