Assay Details
Gene Symbol: MLX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035061.2 | NP_001030233.1 | 3 - 4 | 194 | 66 | |
GenBank mRNA | BC103322.1 | - | 3 - 4 | 197 | 66 | |
BT030524.1 | - | 3 - 4 | 167 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
