Assay Details
Gene Symbol: DARS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035085.1 | NP_001030257.1 | 8 - 9 | 714 | 123 | |
GenBank mRNA | BC103319.1 | - | 8 - 9 | 714 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
