Assay Details
Gene Symbol: ARV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035302.2 | NP_001030379.1 | 4 - 5 | 725 | 94 | |
GenBank mRNA | BC102679.1 | - | 4 - 5 | 726 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
