Assay Details
Gene Symbol: RPL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035306.2 | NP_001030383.1 | 1 | 72 | 75 | |
GenBank mRNA | BC105198.1 | - | 1 | 73 | 75 | |
BT021482.1 | - | 1 - 2 | 49 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC105198.1, NM_001035306.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
