Assay Details
Gene Symbol: NEDD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035333.2 | NP_001030410.1 | 13 - 14 | 1861 | 103 | |
GenBank mRNA | BC107541.1 | - | 13 - 14 | 1862 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
