Assay Details
Gene Symbol: OXA1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035335.2 | NP_001030412.1 | 6 - 7 | 845 | 137 | |
GenBank mRNA | BC103367.1 | - | 6 - 7 | 848 | 137 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
