Assay Details
Gene Symbol: SYNJ2BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035355.2 | NP_001030432.1 | 2 - 3 | 298 | 62 | |
GenBank mRNA | BC102451.1 | - | 2 - 3 | 299 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
