Assay Details
Gene Symbol: SC5D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035356.1 | NP_001030433.1 | 1 - 2 | 64 | 122 | |
GenBank mRNA | BC103338.1 | - | 1 - 2 | 64 | 122 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC103338.1, NM_001035356.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
