Assay Details
Gene Symbol: PPP1R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035392.2 | NP_001030469.1 | 4 - 5 | 699 | 152 | |
GenBank mRNA | BC102669.1 | - | 4 - 5 | 700 | 152 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
