Assay Details
Gene Symbol: ATP1B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035393.2 | NP_001030470.1 | 2 - 3 | 409 | 89 | |
GenBank mRNA | BC102454.1 | - | 2 - 3 | 411 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
