Assay Details
Gene Symbol: LASP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035394.1 | NP_001030471.1 | 3 - 4 | 381 | 67 | |
GenBank mRNA | BC107542.1 | - | 3 - 4 | 381 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
