Assay Details
Gene Symbol: MRPS36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035438.1 | NP_001030515.1 | 3 - 4 | 289 | 117 | |
GenBank mRNA | BC103454.1 | - | 3 - 4 | 289 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
