Assay Details
Gene Symbol: IPMK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035462.1 | NP_001030539.1 | 5 - 6 | 929 | 101 | |
GenBank mRNA | BC102793.1 | - | 5 - 6 | 929 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
