Assay Details
Gene Symbol: CHTOP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035481.2 | NP_001030558.1 | 4 - 5 | 651 | 102 | |
GenBank mRNA | BC103347.1 | - | 4 - 5 | 652 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
