Assay Details
Gene Symbol: UBE2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037449.2 | NP_001032526.1 | 4 - 5 | 479 | 114 | |
GenBank mRNA | BC108195.1 | - | 4 - 5 | 480 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
