Assay Details
Gene Symbol: FCF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037452.1 | NP_001032529.1 | 1 - 2 | 35 | 68 | |
GenBank mRNA | BC108165.1 | - | 1 - 2 | 35 | 68 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC108165.1, NM_001037452.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
