Assay Details
Gene Symbol: FCF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037452.1 | NP_001032529.1 | 5 - 6 | 398 | 91 | |
GenBank mRNA | BC108165.1 | - | 5 - 6 | 398 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
