Assay Details
Gene Symbol: MRPL27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037453.2 | NP_001032530.1 | 3 - 4 | 263 | 106 | |
GenBank mRNA | BC108174.1 | - | 3 - 4 | 264 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
