Assay Details
Gene Symbol: EEF1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037464.2 | NP_001032541.1 | 5 - 6 | 872 | 53 | |
GenBank mRNA | BC108110.1 | - | 5 - 6 | 873 | 53 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
