Assay Details
Gene Symbol: UXT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037471.2 | NP_001032548.1 | 3 - 4 | 209 | 88 | |
GenBank mRNA | BC108205.1 | - | 3 - 4 | 210 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
