Assay Details
Gene Symbol: MDP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037486.2 | NP_001032563.1 | 2 - 3 | 208 | 69 | |
GenBank mRNA | BC108197.1 | - | 2 - 3 | 208 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
