Assay Details
Gene Symbol: MDP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037486.2 | NP_001032563.1 | 4 - 5 | 408 | 116 | |
NM_001164221.1 | NP_001157693.1 | 5 - 6 | 519 | 116 | ||
GenBank mRNA | BC108197.1 | - | 4 - 5 | 408 | 116 | |
EV663472.1 | - | 5 - 6 | 501 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
