Assay Details
Gene Symbol: MRPL42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037622.2 | NP_001032711.1 | 2 - 3 | 123 | 138 | |
GenBank mRNA | BC109670.1 | - | 2 - 3 | 124 | 138 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC109670.1, NM_001037622.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
