Assay Details
Gene Symbol: MRPL42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037622.2 | NP_001032711.1 | 3 - 4 | 288 | 152 | |
GenBank mRNA | BC109670.1 | - | 3 - 4 | 289 | 152 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
