Assay Details
Gene Symbol: SSR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038503.2 | NP_001033592.1 | 2 - 3 | 208 | 102 | |
GenBank mRNA | BC109500.1 | - | 2 - 3 | 209 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
