Assay Details
Gene Symbol: MRPL20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038506.2 | NP_001033595.1 | 1 - 2 | 118 | 79 | |
GenBank mRNA | BC109775.1 | - | 1 - 2 | 119 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
